EMAIL THIS PAGE TO A FRIEND
Organic letters
Expedient synthesis of chiral oxazolidinone scaffolds via rhodium-catalyzed asymmetric ring-opening with sodium cyanate.
PMID 23394120
Abstract
A method for synthesizing chiral oxazolidinone scaffolds from readily available oxabicyclic alkenes is described. The reaction utilizes a domino sequence of Rh(I)-catalyzed asymmetric ring-opening (ARO) with sodium cyanate as a novel nucleophile followed by intramolecular cyclization to generate oxazolidinone products in excellent enantioselectivities (trans stereochemistry).