Schizophrenia research
Identifying clinically meaningful symptom response cut-off values on the SANS in predominant negative symptoms.
PMID 23394742
The treatment and measurement of negative symptoms are currently at issue in schizophrenia, but the clinical meaning of symptom severity and change is unclear. To offer a clinically meaningful interpretation of severity and change scores on the Scale for the Assessment of Negative Symptoms (SANS). Patients were intention-to-treat participants (n=383) in two double-blind randomized placebo-controlled clinical trials that compared amisulpride with placebo for the treatment of predominant negative symptoms. Equipercentile linking was used to examine extrapolation from (a) CGI-S to SANS severity ratings, and (b) CGI-I to SANS percentage change (n=383). Linking was conducted at baseline, 8-14 days, 28-30 days, and 56-60 days of the trials. Across visits, CGI-S ratings of 'not ill' linked to SANS scores of 0-13, and ranged to 'extreme' ratings that linked to SANS scores of 102-105. The relationship between the CGI-S and the SANS severity scores assumed a linear trend (1=0-13, 2=15-56, 3=37-61, 4=49-66, 5=63-75, 6=79-89, 7=102-105). Similarly the relationship between CGI-I ratings and SANS percentage change followed a linear trend. For instance, CGI-I ratings of 'very much improved' were linked to SANS percent changes of -90 to -67, 'much improved' to -50 to -42, and 'minimally improved' to -21 to -13. The current results uniquely contribute to the debate surrounding negative symptoms by providing clinical meaning to SANS severity and change scores and so offer direction regarding clinically meaningful response cut-off scores to guide treatment targets of predominant negative symptoms.
Related Materials
Product #
Molecular Formula
Add to Cart