EMAIL THIS PAGE TO A FRIEND
Seminars in pediatric surgery
Management of prenatally diagnosed lung lesions.
PMID 23395142
Abstract
Prenatal diagnosis provides insight into the in utero evolution of fetal thoracic lesions such as congenital cystic adenomatoid malformation (CCAM), bronchopulmonary sequestration (BPS), or hybrid lesions. Serial sonographic study of fetuses with thoracic lesions has helped define the natural history of these lesions, determine the pathophysiologic features that affect clinical outcome, and formulate in utero and postnatal management based on prognosis.