EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and biological evaluation of 1-(2-hydroxy-3-phenyloxypropyl)piperazine derivatives as T-type calcium channel blockers.
PMID 23395659
Abstract
To obtain selective and potent inhibitor for T-type calcium channel by ligand based drug design, 2-hydroxy-3-phenoxypropyl piperazine derivatives were synthesized and evaluated for in vitro activities. Compound 6m and 6q showed high selectivity over hERG channel (IC50 ratio of hERG/α1G6m=8.5, 6q=18.38) and they were subjected to measure pharmacokinetics profiles. Among them compound 6m showed an excellent pharmacokinetic profile in rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
80621
Piperazine, BioUltra, anhydrous, ≥99.0% (T)
C4H10N2
P7003
Piperazine hexahydrate, analytical standard
C4H10N2 · 6H2O