EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Sensitive amperometric biosensor for phenolic compounds based on graphene-silk peptide/tyrosinase composite nanointerface.
PMID 23395727
Abstract
New graphene-silk peptide (Gr-SP) nanosheets were prepared and successfully fabricated with tyrosinase (Tyr) as a novel biosensor for the determination of phenolic compounds. The Gr-SP nanosheets were fully characterized with transmission electron microscopy, x-ray diffraction, x-ray photoelectron spectroscopy, UV/Vis and FTIR spectra. The developed biosensors were also characterized with scanning electronic microscopy and electrochemical impedance spectroscopy. Using bisphenol A (BPA) as a model substrate in the sensing system, a number of key factors including the volume of Gr-SP-Tyr solution, the applied potential, pH values, temperature, and the Tyr/Gr-SP ratio that influence the analytical performance of the biosensor were investigated. The biosensor gave a linear response on the concentration ranges of 0.001-16.91 μM for catechol with the sensitivity of 7634 mA M(-1)cm(-2), 0.0015-21.12 μM for phenol with the sensitivity of 4082 mA M(-1)cm(-2), and 0.002-5.48 μM for BPA with the sensitivity of 2511 mA M(-1)cm(-2). The low detection limits were estimated to be 0.23, 0.35 and 0.72 nM (S/N=3) for catechol, phenol and BPA, respectively. The biosensors also exhibit good repeatability and long-term stability. The practical application of the biosensor was also demonstrated by the determination of BPA leaching from commercial plastic drinking bottles.