EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
A pH-responsive nano-carrier with mesoporous silica nanoparticles cores and poly(acrylic acid) shell-layers: fabrication, characterization and properties for controlled release of salidroside.
PMID 23395877
Abstract
A novel pH-responsive nano-carrier MSNs-PAA, possessing mesoporous silica nanoparticles (MSNs) cores and poly(acrylic acid) (PAA) shell-layers, was developed for controlled release of salidroside. The vinyl double bonds modified MSNs were synthesized by using cetyltrimethylammonium bromide (CTAB) as templates, tetraethyl orthosilicate (TEOS) as silicon source, and 3-(trimethoxylsilyl) propyl methacrylate (MPS) as surface modification functionalities. The pH-responsive layers of PAA were grafted onto the vinyl double bonds of the MSNs via precipitation polymerization, producing the MSNs-PAA with a hollow cubic core and mesoporous shell with penetrating pore channels. The characteristic results also showed that PAA was successfully grafted onto the surface of the MSNs. The MSNs-PAA was investigated as carriers for loading and regulating the release of salidroside in different pH solutions for the first time. The results demonstrated that the PAA layers on the surface of MSNs-PAA exhibited opened and closed states at different pH values, and thus could regulate the uptake and release of salidroside. The application of such pH-responsive nano-carrier might offer a potential platform for controlled delivery and increasing the bioavailability of drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
292737
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
C19H42ClN
374598
Cetyltrimethylammonium hydrogensulfate, 99%
C19H43NO4S
52371
Hexadecyltrimethylammonium bisulfate, for ion pair chromatography, LiChropur, ≥99.0% (T)
C19H43NO4S
52366
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
C19H42ClN
439231
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
C19H43NO
52387
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
C19H43NO
447013
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
420344
Poly(acrylic acid sodium salt), average Mw ~2,100
81117
Poly(acrylic acid sodium salt), analytical standard, for GPC, 1,250
81123
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000
81131
Poly(acrylic acid sodium salt), analytical standard, for GPC, 83,000
323667
Poly(acrylic acid)
181285
Poly(acrylic acid), average Mv ~450,000
306215
Poly(acrylic acid), average Mv ~1,250,000
306231
Poly(acrylic acid), average Mv ~4,000,000
306223
Poly(acrylic acid), average Mv ~3,000,000
181293
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
416002
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
523925
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
535931
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
416029
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
416010
Poly(acrylic acid, sodium salt) solution, average Mw ~1,200, 45 wt. % in H2O
43866
Salidroside, analytical standard
C14H20O7