EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Syntheses, crystal structures and photophysical properties of Ag(I) coordination complexes.
PMID 23396006
Abstract
Three Ag(I) coordination complexes, [Ag(pyzca)]n (1) (Hpyzca=pyrazine-2-carboxylic acid), Ag[(HNA)(NA)(H2O)4] (2) (HNA=nicotinic acid) and [Ag1Ag2-(2,3-pydc)]n (3) (H2pydc=pyridine-2, 3-dicarboxylic acid) have been synthesized by the hydrothermal method. Their structures are determined by single-crystal X-ray diffractometer. Complex 1 possesses the 3D infinite structure. Complex 2 is a mononuclear coordination complex and exhibits one 3D infinite network by a large number of hydrogen bonds. In complex 3, two crystallographic independent Ag(I) ions exist, i.e., Ag1 is three-coordinate and Ag2 is two-coordinate, forming the 2D layer structure. At room temperature in the solid state, the IR, FP and UV-Vis spectra of all complexes were measured and investigated in detail. The emission and absorption spectra of the three complexes and their corresponding ligands are compared and analyzed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
170860
Nicotinic acid sodium salt, 98%
C6H4NNaO2