EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Total synthesis of kottamide E.
PMID 23396320
Abstract
The first synthesis of kottamide E, a marine natural product containing a 5,6-dibromoindole linked via a (Z)-enamide to an unusual 1,2-dithiolane-containing amino acid, is reported.