EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Multifunctionalization of alkenes via aerobic oxynitration and sp3 C-H oxidation.
PMID 23396373
Abstract
A method for direct functionalization of three positions including an unactivated C-H bond of aliphatic alkenes using tert-butyl nitrite and molecular oxygen to give γ-lactols has been developed. The present reaction proceeds through a sequence of radical processes involving oxynitration followed by aerobic oxidation of an sp(3) C-H bond. This multifunctionalization reaction requires neither metallic reagents nor photolysis and proceeds under mild conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226637
Butyl nitrite, 95%
C4H9NO2