EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Three-dimensional homochiral manganese-lanthanide frameworks based on chiral camphorates with multi-coordination modes.
PMID 23396508
Abstract
A series of 3D homochiral manganese-lanthanide frameworks have been synthesized based on chiral camphoric acid. In the heterometallic features, D-camphoric acids are unprecedentedly embedded in three coordination modes.