EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Multi-responsive nitrobenzene-based amphiphilic random copolymer assemblies.
PMID 23396576
Abstract
A photo-, acid- and thermo-responsive nitrobenzene-based amphiphilic copolymer was synthesized and the corresponding multi-responsive behavior of the copolymer assemblies was revealed by TEM, DLS and AFM. The morphological changes of the assemblies under multi-stimuli endowed them with a controlled release of encapsulated molecules.