EMAIL THIS PAGE TO A FRIEND
Journal of investigational allergology & clinical immunology
Nonhistaminergic idiopathic angioedema: clinical response to icatibant.
PMID 23397675
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3259
Bradykinin acetate salt, powder, ≥98% (HPLC)
C50H73N15O11 · xC2H4O2
H157
HOE 140, ≥94%
C59H89N19O13S