EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Late-stage deoxyfluorination of alcohols with PhenoFluor.
PMID 23397884
Abstract
An operationally simple protocol for the selective deoxyfluorination of structurally complex alcohols is presented. Several fluorinated derivatives of natural products and pharmaceuticals have been prepared to showcase the potential of the method for late-stage diversification and its functional group compatibility. A series of simple guidelines for predicting the selectivity in substrates with multiple alcohols is given.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
795291
Phenofluor solution, 0.1 M in toluene
C27H36F2N2