EMAIL THIS PAGE TO A FRIEND
Infectious disease clinics of North America
What is the role of respiratory viruses in community-acquired pneumonia?: What is the best therapy for influenza and other viral causes of community-acquired pneumonia?
PMID 23398872
Abstract
Respiratory viruses have long been appreciated as a cause of community acquired pneumonia (CAP), particularly among children, people with serious medical comorbidities, and military recruits. They are increasingly recognized as a cause of CAP among adults. Polymerase chain reaction-based testing has allowed detection of newer agents and improved the ability to detect such viral infections as influenza virus and rhinovirus. Coinfection with viruses and bacteria is common and it remains challenging to determine which patients have only viral infection as the cause of CAP. Better ways to diagnose viral CAP and to integrate detection into management, and better treatment options for noninfluenza respiratory viral infections are needed.