Carbohydrate polymers
Surface grafting of Corchorus olitorius fibre: a green approach for the development of activated bioadsorbent.
PMID 23399266
The present work is an endeavor to prepare lignocellulosic biomass based adsorbent, suitable for removal of organic and inorganic pollutants from industrial effluents. Lignocellulosic Corchorus olitorius fibre (jute fibre) surface was grafted with naturally available polyphenol, tannin, preceded by the epoxy-activation of fibre surface with epichlorohydrin under mild condition in an aqueous suspension. The reaction parameters for the modification, viz., concentration of epichlorohydrin and tannin, time, and temperature were optimized. The successful occurrence of surface modification of jute fibre (JF) was characterized and estimated from weight gain percent, elemental analysis, Fourier transform infrared spectroscopy (FTIR), X-ray diffraction, scanning electron and atomic force microscopy, and thermogravimetric analysis. An extensive analysis of deconvoluted FTIR spectra using the Voigt model was utilized to ensure the surface grafting. The microbiological susceptibility study revealed high persistency of JF towards biodegradation after efficient grafting with tannin.