EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Illicit drugs: What dermatologists need to know.
PMID 23399461
Abstract
We review the most common systemic and cutaneous signs of heroin, cocaine, methamphetamine, Ecstasy, and marijuana use. We also provide an overview of the skin and soft-tissue infections frequently found in intravenous drug users and the effects of the adulterants added to the drugs.