EMAIL THIS PAGE TO A FRIEND
Experimental neurology
Oligodendroglial alpha-synucleinopathy and MSA-like cardiovascular autonomic failure: experimental evidence.
PMID 23399889
Abstract
Multiple system atrophy (MSA) is a fatal, rapidly progressive neurodegenerative disease with limited symptomatic treatment options. Discrimination of MSA from other degenerative disorders crucially depends on the presence of early and severe cardiovascular autonomic failure (CAF). We have previously shown that neuropathologic lesions in the central autonomic nuclei similar to the human disease are present in transgenic MSA mice generated by targeted oligodendroglial overexpression of α-syn using the PLP promoter. We here explore whether such lesions result in abnormalities of heart rate variability (HRV) and circadian rhythmicity which are typically impaired in MSA patients. HRV analysis was performed in five month old transgenic PLP-α-syn (tg) MSA mice and age-matched wild type controls. Decreased HRV and alterations in the circadian rhythmicity were detected in the tg MSA group. The number of choline-acetyltransferase-immunoreactive neurons in the nucleus ambiguus was significantly decreased in the tg group, whereas the levels of arginine-vasopressin neurons in the suprachiasmatic and paraventricular nucleus were not affected. Our finding of impaired HRV and circadian rhythmicity in tg MSA mice associated with degeneration of the nucleus ambiguus suggests that a cardinal non-motor feature of human MSA can be reproduced in the mouse model strengthening its role as a valuable testbed for studying selective vulnerability and assessing translational therapies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V0377
[Arg8]-Vasopressin solution, Grade VI (synthetic), ~100 I.U./mL in 0.9% NaCl
C46H65N15O12S2