EMAIL THIS PAGE TO A FRIEND
Chemical Society reviews
Chromogenic and fluorogenic chemosensors and reagents for anions. A comprehensive review of the years 2010-2011.
PMID 23400370
Abstract
This review focuses on examples reported in the years 2010-2011 dealing with the design of chromogenic and fluorogenic chemosensors or reagents for anions.