EMAIL THIS PAGE TO A FRIEND
Arthritis and rheumatism
Developing better biomarkers for connective tissue disease-associated interstitial lung disease: citrullinated hsp90 autoantibodies in rheumatoid arthritis.
PMID 23400799