EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia gastroenterologiia = Experimental & clinical gastroenterology
[Using ursodez in the treatment of refractory form of gastroesophageal reflux disease].
PMID 23402158