EMAIL THIS PAGE TO A FRIEND
Ugeskrift for laeger
[Possible clinical potential in reverting muscular block with sugammadex in anaesthesia and surgery].
PMID 23402253
Abstract
Neuromuscular blockers (NMBs) provide good conditions for endotracheal intubation and surgery. NMBs have been associated with higher morbidity and mortality, mainly due to post-operative residual neuromuscular block. This may become history with the advent of sugammadex - an antidote to the NMB rocuronium - which within 1-3 minutes neutralizes the effects of rocuronium. High-dose rocuronium is now an alternative to suxamethonium in acute or short procedures and in a situation, where ventilation/intubating cannot be performed, sugammadex can reverse the rocuronium blockade within minutes.