EMAIL THIS PAGE TO A FRIEND
Functional neurology
Continuous intestinal infusion of levodopa/carbidopa in advanced Parkinson's disease: efficacy, safety and patient selection.
PMID 23402675
Abstract
Long-term oral therapy with levodopa is associated with the development of motor fluctuations and dyskinesia in a large percentage of patients with Parkinson's disease (PD). Motor complications are associated with a number of non-motor symptoms and have a negative impact on disability and quality of life. There are three therapeutic options available for the management of patients at this advanced stage: high frequency deep brain stimulation, continuous subcutaneous infusion of apomorphine, and continuous intestinal infusion of levodopa/carbidopa. On the basis of published data and in consideration of the risk-benefit profile of current therapeutic strategies, we here propose an algorithm to help clinicians select the most suitable treatment option for patients with advanced PD.