EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Neuroprotective effects of mercaptoethylleonurine and mercaptoethylguanidine analogs on hydrogen peroxide-induced apoptosis in human neuronal SH-SY5Y cells.
PMID 23403085
Abstract
A series of mercaptoethylleonurine and mercaptoethylguanidine derivatives were designed and synthesized. Their neuroprotective effects toward H2O2-induced apoptosis were investigated in human SH-SY5Y cells. The results from these studies identified several potent compounds, with compound 8k emerging as the most effective. Further investigation demonstrated that 8k reduced H2O2-induced activation of mitochondrial apoptosis by inhibiting the expression of Bax and elevating the expression of Bcl-2. Moreover, the molecular mechanism underlying the observed neuroprotective effects of 8k was exerted via the Akt and JNK pathways. Compound 8k can be a lead compound for further discovery of neuroprotective medicine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9940
Mercaptoethylguanidine hemisulfate salt, ≥98%
C3H9N3S · 0.5H2SO4