EMAIL THIS PAGE TO A FRIEND
Gene
Physical mapping and cloning of RAD56.
PMID 23403232
Abstract
Here we report the physical mapping of the rad56-1 mutation to the NAT3 gene, which encodes the catalytic subunit of the NatB N-terminal acetyltransferase in Saccharomyces cerevisiae. Mutation of RAD56 causes sensitivity to X-rays, methyl methanesulfonate, zeocin, camptothecin and hydroxyurea, but not to UV light, suggesting that N-terminal acetylation of specific DNA repair proteins is important for efficient DNA repair.