EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A macrocyclic aromatic pyridone pentamer as a highly efficient organocatalyst for the direct arylations of unactivated arenes.
PMID 23403921
Abstract
A macrocyclic aromatic pyridone pentamer was shown to catalyze highly efficient transition-metal-free arylations of unactivated aromatic C-H bonds with aryl iodides and bromides in the presence of potassium tert-butoxide.