PloS one
Biochemical similarities and differences between the catalytic [4Fe-4S] cluster containing fumarases FumA and FumB from Escherichia coli.
PMID 23405168
Abstract
The highly homologous [4Fe-4S] containing fumarases FumA and FumB, sharing 90% amino acid sequence identity, from Escherichia coli are differentially regulated, which suggests a difference in their physiological function. The ratio of FumB over FumA expression levels increases by one to two orders of magnitude upon change from aerobic to anaerobic growth conditions. To understand this difference in terms of structure-function relations, catalytic and thermodynamic properties were determined for the two enzymes obtained from homologous overexpression systems. FumA and FumB are essentially identical in their Michaelis-Menten kinetics of the reversible fumarate to L-malate conversion; however, FumB has a significantly greater catalytic efficiency for the conversion of D-tartrate to oxaloacetate consistent with the requirement of the fumB gene for growth on D-tartrate. Reduction potentials of the [4Fe-4S](2+) Lewis acid active centre were determined in mediated bulk titrations in the presence of added substrate and were found to be approximately -290 mV for both FumA and FumB. This study contradicts previously published claims that FumA and FumB exhibit different catalytic preferences for the natural substrates L-malate and fumarate. FumA and FumB differ significantly only in the catalytic efficiency for the conversion of D-tartrate, a supposedly non-natural substrate. The reduction potential of the substrate-bound [4Fe-4S] active centre is, contrary to previously reported values, close to the cellular redox potential.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T206
D-(−)-Tartaric acid, ReagentPlus®, 99%
C4H6O6
483796
D-(−)-Tartaric acid, 99%
C4H6O6
95320
D-(−)-Tartaric acid, puriss., unnatural form, ≥99.0% (T)
C4H6O6
03918
DL-Tartaric acid, anhydrous, for ion chromatography, ≥99.5% (T)
C4H6O6
T400
DL-Tartaric acid, ReagentPlus®, 99%
C4H6O6
W304401
DL-Tartaric acid, ≥99%
C4H6O6
F1757 Fumarase from porcine heart, ammonium sulfate suspension, 300-500 units/mg protein (biuret)
251380
L-(+)-Tartaric acid, ACS reagent, ≥99.5%
C4H6O6
W304409
L-(+)-Tartaric acid, ≥99.7%, FCC, FG
C4H6O6
T109
L-(+)-Tartaric acid, ≥99.5%
C4H6O6
T1807
L-(+)-Tartaric acid, BioXtra
C4H6O6
33801
L-(+)-Tartaric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)
C4H6O6
41447
L-(+)-Tartaric acid, certified reference material, TraceCERT®
C4H6O6
27506
L-(+)-Tartaric acid, puriss., meets analytical specification of Ph. Eur., NF, 99.7-100.5% (calc. to the dried substance), powder
C4H6O6
95318
L-(+)-Tartaric acid, tested according to Ph.Eur.
C4H6O6
798665
L-(+)-Tartaric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
C4H6O6
O4126
Oxaloacetic acid, ≥97% (HPLC)
C4H4O5
O7753
Oxaloacetic acid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)
C4H4O5
O9504
Oxaloacetic acid, Hybri-Max, powder, suitable for hybridoma
C4H4O5