EMAIL THIS PAGE TO A FRIEND
The Journal of family practice
Clinical Inquiry: which treatments help women with reduced libido?
PMID 23405381
Abstract
Several treatments produce modest, but statistically significant, clinical increases in sexual desire and function in women. The testosterone transdermal patch improves hypoactive sexual desire disorder (HSDD) in postmenopausal women.