EMAIL THIS PAGE TO A FRIEND
Organic letters
Structures and biomimetic synthesis of novel α-pyrone polyketides of an endophytic Penicillium sp. in Catharanthus roseus.
PMID 23405967
Abstract
Novel polyketides, citreoviripyrone A (1) and B (2), known citreomontanin (3), and (-)-citreoviridin (4) were isolated from the mycelium of the endophytic fungus. The endophytic fungus, which belongs to the genus Penicillium, was separated from surface-sterilized healthy leaves of Catharanthus roseus. The structures of 1 and 2 were determined on the basis of NMR data, and 1 was characterized as an α-pyrone polyketide featuring bicyclo[4.2.0]octadiene. The biomimetic synthesis of 1 and 2 from 3 elucidated a plausible biosynthetic pathway. Both Zn(II)-type and NAD(+)-dependent histone deacetylase inhibitors significantly enhanced the production of 1 and 3.