Toxicology and industrial health
The potent effects of ginseng root extract and memantine on cognitive dysfunction in male albino rats.
PMID 23406953
The study determined the maximum intraperitoneal (ip) scopolamine dose inducing memory impairment in rats (2 mg/kg) compared to 0.5 or 1 mg/kg dose. The effect reflected by significant increase from normal in the latency time required for rats to find the hidden platform in water maze task and acetylcholinesterase (AChE) activities in cortex, hippocampus and striatum. The dose-related histopathological effect via the hemorrhage, vacuolation and gliosis in cortex and hippocampus is assessed. Then the study investigated the potency of Panax ginseng root extract on scopolamine cognitive dysfunction rat model compared to memantine hydrochloride as reference Food and Drug Administration approved. Ginseng extract was administered at dose 100 or 200 mg/kg/day and memantine at 20 mg/kg/day orally for 2 weeks. All treatments showed improvement in the water maze task, however, ginseng (200 mg/kg) group acquired the advantage without statistical difference control. Scopolamine (2 mg/kg ip) group showed significant increase in AChE reactivity and glutamate level and reduced monoamines (norepinephrine, dopamine and serotonin) and γ-aminobutyric acid contents in cortex, hippocampus and striatum. Ginseng extract in a dose-dependent manner appears effective as memantine and can improve memory impairment through the retrieved homeostasis via neurotransmitter levels and AChE activities in rat brain areas with partial effect on the histological feature of the brain tissue.
Related Materials
Product #
Molecular Formula
Add to Cart
(−)-Scopolamine hydrobromide trihydrate, ≥98% (HPLC), powder
C17H21NO4 · HBr · 3H2O
Scopolamine hydrobromide, meets USP testing specifications
C17H21NO4 · HBr · 3H2O