EMAIL THIS PAGE TO A FRIEND
The Journal of infectious diseases
Tuberculosis relapse in Vietnam is significantly associated with Mycobacterium tuberculosis Beijing genotype infections.
PMID 23408848
Abstract
In Vietnam, the Mycobacterium tuberculosis Beijing genotype is associated with multi-drug resistance and is emerging. A possible explanation for this genotype's success is an increased rate of relapse. In a prospective cohort study, isolates from patients with smear-positive tuberculosis were subjected to drug susceptibility testing and to spoligotyping and variable number of tandem repeats typing before treatment and after recurrence of tuberculosis. Among 1068 patients who were actively followed up over 18 months for recurrence, 23 relapse cases occurred (1.39 cases/100 person-years). After adjustment for genotype, tuberculosis treatment history, and drug resistance, relapse was significantly associated with the Beijing genotype (adjusted hazard ratio [aHR], 5.48; 95% confidence interval [CI], 2.06-14.55) and isoniazid resistance (aHR, 5.91; 95% CI, 2.16-16.16). The strongly increased relapse rate in tuberculosis cases caused by Beijing strains probably contributes to the successful spread of this genotype in Vietnam and elsewhere.