EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Metal-organic frameworks as hosts for photochromic guest molecules.
PMID 23409796
Abstract
Several metal-organic framework compounds (MOF-5, MIL-68(Ga), MIL-68(In), MIL-53(Al)) were loaded with azobenzene (AZB), as confirmed by XRPD measurements and elemental analysis. By IR spectroscopy, it was shown that the light-induced trans/cis isomerization of AZB in these hybrid host-guest compounds is improved compared to that of solid AZB. A population of the excited cis state up to 30% has been obtained for AZB0.66@MIL-68(In). However, no light-induced trans/cis isomerization was observed for AZB0.5@MIL-53(Al). Structural models obtained from high-resolution synchrotron powder diffraction data show that AZB molecules are densely packed within the channels of MIL-53(Al) so that no trans/cis isomerization can occur. A different situation was observed for AZB in the larger channels of MIL-68(Ga). Thus, this investigation shows the influence of the host material on the switching behavior of the embedded AZB molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
424633
Azobenzene, 98%
C12H10N2
47959 Azobenzene solution, 2000 μg/mL in methanol, analytical standard
C12H10N2
203319
Gallium, 99.9995% trace metals basis
Ga
263273
Gallium, 99.999% trace metals basis
Ga
263265
Gallium, 99.99% trace metals basis
Ga
772631
Gallium, pieces, 99.99999% trace metals basis
Ga
57083
Indium, bars, 99.95%
In
264113
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
In
326607
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
In
357278
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
In
326631
Indium, foil, thickness 0.5 mm, 99.999% trace metals basis
In
357286
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
In
264040
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
In
357294
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
In
264059
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
In
357308
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
In
326615
Indium, pieces, 99.99% trace metals basis
In
203432
Indium, powder, −60 mesh, 99.999% trace metals basis
In
264032
Indium, powder, −100 mesh, 99.99% trace metals basis
In
277959
Indium, powder, 99.99% trace metals basis
In
385379
Indium, rod, diam. 10 mm, 99.9998% trace metals basis
In
264091
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
In
357065
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
In
340863
Indium, wire, diam. 1.5 mm, 99.99% trace metals basis
In
357073
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
In
264075
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
In
264067
Indium, wire, diam. 0.5 mm, 99.995% trace metals basis
In
326623
Indium, wire, diam. 0.25 mm, 99.999% trace metals basis
In
442465
Azobenzene, analytical standard
C12H10N2