EMAIL THIS PAGE TO A FRIEND
Organic letters
Cascade synthesis of fenpiprane and related pharmaceuticals via rhodium-catalyzed hydroaminomethylation.
PMID 23410073
Abstract
A novel rhodium catalytic system with Naphos as ligand was developed for an efficient hydroaminomethylation of 1,1-diphenylethene under relatively mild conditions. This will allow for an atom-economic and environmentally benign synthesis of fenpiprane and related pharmaceuticals.