EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of diphenylhexatriene by the Pd-catalyzed dimerization of cinnamyl acetate.
PMID 23414259
Abstract
A mild and operationally simple method to synthesize diphenylhexatriene (DPH) is reported. The method relies on the Pd-catalyzed dimerization of cinnamyl acetate and provides efficient access to DPH in a single step.