EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Virtual colonoscopy: stamp of approval or word of warning?
PMID 23414649
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
243353
Barium sulfate, ReagentPlus®, 99%
BaO4S