EMAIL THIS PAGE TO A FRIEND
Journal of clinical lipidology
Adverse reactions of Achilles tendon xanthomas in three hypercholesterolemic patients after treatment intensification with niacin and bile acid sequestrants.
PMID 23415439
Abstract
Multiple cholesterol-reducing therapies have been shown to induce the regression of tendon xanthoma in patients with familial hypercholesterolemia. We present 3 cases of adverse reactions in Achilles tendon xanthomas after the addition of niacin and bile acid sequestrants to ongoing statin therapy. Reduction in tendon dimensions and marked softening of xanthomas were interpreted as cholesterol removal from heavily infiltrated tissue sites. In 2 cases, changes in the xanthomas occurred despite only minor lipoprotein improvements, raising the possibility of direct drug effects in cholesterol-infiltrated tissue. Intriguingly, recent studies have described niacin receptor-mediated effects in macrophages. In summary, although adverse reactions in Achilles tendon xanthomas appear to be infrequent, clinicians should be aware of this phenomenon in their patients after intensifying lipid treatments, especially with the use of niacin in patients with familial hypercholesterolemia. Xanthoma responses may provide clues to new pharmacologic effects in cholesterol-infiltrated tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
170860
Nicotinic acid sodium salt, 98%
C6H4NNaO2