EMAIL THIS PAGE TO A FRIEND
European journal of cancer (Oxford, England : 1990)
Excess breast cancer risk in first degree relatives of CHEK2∗1100delC positive familial breast cancer cases.
PMID 23415889
Abstract
The CHEK2∗1100delC mutation confers a relative risk of two for breast cancer (BC) in the general population. This study aims to explore the excess cancer risk due to the CHEK2∗1100delC mutation within a familial non-BRCA1/2 breast cancer setting. Cancer incidences were compared between first degree relatives of 107 familial breast cancer patients positive for the CHEK2∗1100delC mutation (CHEK2 positive families) and first degree relatives of 314 familial breast cancer patients without the CHEK2∗1100delC mutation (CHEK2 negative families). All families were derived from the same pool of familial non-BRCA1/2 breast cancer families (n=2554). Medical information of 2188 first degree relatives of these families was analysed for cancer risk. CHEK2∗1100delC status of relatives was unknown. Increased breast cancer risk (hazard ratio (HR) 2.0 (95% confidence interval (CI): 1.4-2.7), p<0.001) was observed in sisters of CHEK2∗1100delC positive index cases compared to sisters of CHEK2∗1100delC negative index cases. HR was 1.6 (95% CI: 1.0-2.4) for mothers of CHEK2 positive versus negative index cases (p=0.041). For second primary breast cancers HR was increased in CHEK2∗1100delC positive index cases (HR 2.1, 95% CI: 1.3-3.3, p=0.003) and their sisters (HR 2.6, 95% CI: 1.1-6.1, p=0.025). There is an excess breast cancer risk in first degree relatives of CHEK2∗1100delC positive non-BRCA1/2 familial breast cancer patients compared to non-CHEK2∗1100delC familial breast cancer relatives. Genotyping for the CHEK2∗1100delC mutation in a familial breast cancer setting contributes to optimal clinical surveillance in countries in which this mutation is prevalent. Carriers and female relatives are eligible for stringent breast surveillance programs.