EMAIL THIS PAGE TO A FRIEND
Nature
Non-optimal codon usage affects expression, structure and function of clock protein FRQ.
PMID 23417067
Abstract
Codon-usage bias has been observed in almost all genomes and is thought to result from selection for efficient and accurate translation of highly expressed genes. Codon usage is also implicated in the control of transcription, splicing and RNA structure. Many genes exhibit little codon-usage bias, which is thought to reflect a lack of selection for messenger RNA translation. Alternatively, however, non-optimal codon usage may be of biological importance. The rhythmic expression and the proper function of the Neurospora FREQUENCY (FRQ) protein are essential for circadian clock function. Here we show that, unlike most genes in Neurospora, frq exhibits non-optimal codon usage across its entire open reading frame. Optimization of frq codon usage abolishes both overt and molecular circadian rhythms. Codon optimization not only increases FRQ levels but, unexpectedly, also results in conformational changes in FRQ protein, altered FRQ phosphorylation profile and stability, and impaired functions in the circadian feedback loops. These results indicate that non-optimal codon usage of frq is essential for its circadian clock function. Our study provides an example of how non-optimal codon usage functions to regulate protein expression and to achieve optimal protein structure and function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6763 Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
T8003
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
T1005
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
T8658
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
T7309
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
T9935
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
T8802
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
T9201
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
T6424 Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
T0303 Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
T4799 Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
T7409 Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
T5266 Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
T7168 Trypsin from porcine pancreas, tablet, 1 mg tablet
93614 Trypsin from porcine pancreas, ≥10000 U/mg
93615 Trypsin from porcine pancreas, ~1500 U/mg
85450C Trypsin Powder, Porcine 1:250, Gamma irradiated by SER-TAIN Process, dry powder
T3568 TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)