EMAIL THIS PAGE TO A FRIEND
Radiology
Three-dimensional and multienergy gamma-ray simultaneous imaging by using a Si/CdTe Compton camera.
PMID 23418002
Abstract
To develop a silicon (Si) and cadmium telluride (CdTe) imaging Compton camera for biomedical application on the basis of technologies used for astrophysical observation and to test its capacity to perform three-dimensional (3D) imaging. All animal experiments were performed according to the Animal Care and Experimentation Committee (Gunma University, Maebashi, Japan). Flourine 18 fluorodeoxyglucose (FDG), iodine 131 ((131)I) methylnorcholestenol, and gallium 67 ((67)Ga) citrate, separately compacted into micro tubes, were inserted subcutaneously into a Wistar rat, and the distribution of the radioisotope compounds was determined with 3D imaging by using the Compton camera after the rat was sacrificed (ex vivo model). In a separate experiment, indium 111((111)In) chloride and (131)I-methylnorcholestenol were injected into a rat intravenously, and copper 64 ((64)Cu) chloride was administered into the stomach orally just before imaging. The isotope distributions were determined with 3D imaging after sacrifice by means of the list-mode-expectation-maximizing-maximum-likelihood method. The Si/CdTe Compton camera demonstrated its 3D multinuclear imaging capability by separating out the distributions of FDG, (131)I-methylnorcholestenol, and (67)Ga-citrate clearly in a test-tube-implanted ex vivo model. In the more physiologic model with tail vein injection prior to sacrifice, the distributions of (131)I-methylnorcholestenol and (64)Cu-chloride were demonstrated with 3D imaging, and the difference in distribution of the two isotopes was successfully imaged although the accumulation on the image of (111)In-chloride was difficult to visualize because of blurring at the low-energy region. The Si/CdTe Compton camera clearly resolved the distribution of multiple isotopes in 3D imaging and simultaneously in the ex vivo model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
256544
Cadmium telluride, powder, <250 μm, deposition grade, ≥99.98% trace metals basis
CdTe
716669
Cadmium telluride, 99.9999% trace metals basis
CdTe
307483
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Cl2Cu · 2H2O
221783
Copper(II) chloride dihydrate, reagent grade
Cl2Cu · 2H2O
C3279
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Cl2Cu · 2H2O
467847
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Cl2Cu · 2H2O
459097
Copper(II) chloride dihydrate, 99.999%
Cl2Cu · 2H2O
203319
Gallium, 99.9995% trace metals basis
Ga
263273
Gallium, 99.999% trace metals basis
Ga
772631
Gallium, pieces, 99.99999% trace metals basis
Ga
277959
Indium, powder, 99.99% trace metals basis
In
264032
Indium, powder, −100 mesh, 99.99% trace metals basis
In
264075
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
In
264113
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
In
326607
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
In
357308
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
In
203432
Indium, powder, −60 mesh, 99.999% trace metals basis
In
357073
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
In
264059
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
In
357294
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
In
326615
Indium, pieces, 99.99% trace metals basis
In
264040
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
In
57083
Indium, bars, 99.95%
In
357278
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
In
357065
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
In
357286
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
In
264067
Indium, wire, diam. 0.5 mm, 99.995% trace metals basis
In
264091
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
In
326623
Indium, wire, diam. 0.25 mm, 99.999% trace metals basis
In
340863
Indium, wire, diam. 1.5 mm, 99.99% trace metals basis
In