EMAIL THIS PAGE TO A FRIEND
Catheterization and cardiovascular interventions : official journal of the Society for Cardiac Angiography & Interventions
Nitroprusside fractional flow reserve: hyperemia for less?
PMID 23418119