EMAIL THIS PAGE TO A FRIEND
Clinical otolaryngology : official journal of ENT-UK ; official journal of Netherlands Society for Oto-Rhino-Laryngology & Cervico-Facial Surgery
Up-to-date expert opinion referencing the best evidence available on intraoperative cerebrospinal fluid leak during endoscopic sinus surgery.
PMID 23418966