EMAIL THIS PAGE TO A FRIEND
Haemophilia : the official journal of the World Federation of Hemophilia
History of prophylaxis.
PMID 23419107
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0806 Factor IX human, aqueous glycerol solution, ≥145 units/mg protein