EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Iridium-catalyzed C-H borylation of cyclopropanes.
PMID 23421575
Abstract
The borylation of cyclopropanes catalyzed by the combination of (η(6)-mes)IrBpin3 or [Ir(COD)OMe]2 and a phenanthroline derivative is reported. The borylation occurs selectively at the methylene C-H bonds of the cyclopropane ring over methine or methyl C-H bonds. High diasteroselectivities were observed from reactions catalyzed by the combination of iridium and 2,9-Me2phenanthroline. The cyclopropylboronate esters that are generated are versatile synthetic intermediates that can be converted to trifluoroborate salts, boronic acids, cyclopropylarenes, cyclopropylamines, and cyclopropanols.