EMAIL THIS PAGE TO A FRIEND
Journal of clinical psychopharmacology
Central nervous system-impairing effects of hydroxyzine as a function of histamine availability.
PMID 23422368
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H8885
Hydroxyzine dihydrochloride, ≥98%
C21H27ClN2O2 · 2HCl