EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Synthesis of optically enriched spirocyclic benzofuran-2-ones by bifunctional thiourea-base catalyzed double-Michael addition of benzofuran-2-ones to dienones.
PMID 23423721
Abstract
A highly enantioselective catalytic double-Michael addition reaction of substituted benzofuran-2-ones with divinyl ketones promoted by readily accessible tertiary amine-thiourea Cinchona alkaloids has been developed. A number of optically enriched spirocyclic benzofuran-2-ones were prepared in very good yields (up to 99 %), diastereoselectivities (up to 19:1 d.r.), and very good enantioselectivities (up to 92 % ee). Density functional theory (DFT) calculations were performed to investigate the origin of stereoselectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ALD00382
1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one
C19H12F6O