EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Preparation and evaluation of benzyl methacrylate monoliths for capillary chromatography.
PMID 23424040
Abstract
This paper describes the comprehensive fabrication of monolithic materials for use as stationary phases in capillary liquid chromatography. Several columns were synthesized in the confines of 320 µm i.d. fused-silica capillaries by single-step in situ copolymerization of benzyl methacrylate and ethylene dimethacrylate (EDMA). The polymerization procedure was optimized by varying the reaction time within the range of 0.5-20 h, and by changing the composition contents of the polymeric mixture. The EDMA content showed a predominant influence on the characteristics of the columns and hence, on their chromatographic properties. The optimum value of the thermal initiator corresponded to 5 mg/mL. Changes of the porous, hydrodynamic properties and morphology of the prepared columns were thoroughly investigated and characterized. Different solvents were used as the mobile phase to demonstrate that the resulting monoliths exhibited good permeability and mechanical stability, whereas swelling and shrinking behaviors were observed and discussed. The efficiency and performance toward different sets of analytes were obtained; mixtures of aromatic hydrocarbons and phenolic compounds were successfully separated and evaluated, and adding tetrahydrofuran to the mobile phase showed improvement in both resolution and peak shapes. The characteristics of the columns were also checked in terms of repeatability and reproducibility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
240559
1,4-Butanediol, ReagentPlus®, ≥99%
C4H10O2
493732
1,4-Butanediol, ReagentPlus®, 99%
C4H10O2
96566
1-Propanol, analytical standard
C3H8O
1570406
1-Propanol, United States Pharmacopeia (USP) Reference Standard
C3H8O
PHR1208
1-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O
70448
2-Naphthol, BioXtra, ≥99.0% (GC)
C10H8O
70450
2-Naphthol, fluorescence indicator, ≥99.0%
C10H8O
185507
2-Naphthol, 99%
C10H8O
130109
2-Naphthol, 98%
C10H8O
440159
3-(Trimethoxysilyl)propyl methacrylate, 98%
C10H20O5Si
35837
4-Aminophenol, PESTANAL®, analytical standard
C6H7NO
60034
4-Aminophenol, ≥99% (HPLC)
C6H7NO
A71328
4-Aminophenol, ≥98%
C6H7NO
1021204
4-Aminophenol, United States Pharmacopeia (USP) Reference Standard
C6H7NO
PHR1148
4-Aminophenol (Acetaminophen Related Compound K) (Paracetamol Impurity K), Pharmaceutical Secondary Standard; Certified Reference Material
C6H7NO
07671
Anthracene, certified reference material, TraceCERT®
C14H10
31581
Anthracene, analytical standard
C14H10
48567
Anthracene, analytical standard
C14H10
40076
Anthracene solution, certified reference material, 1000 μg/mL in acetone
C14H10
48647
Anthracene solution, certified reference material, 200 μg/mL in methanol
C14H10
01422
Melting point standard 79-81°C, analytical standard
C10H8
Y0001563
Mesalazine impurity A, European Pharmacopoeia (EP) Reference Standard
C6H7NO
84679
Naphthalene, analytical standard
C10H8
91489
Naphthalene, certified reference material, TraceCERT®
C10H8
PHR1275
Naphthalene, Pharmaceutical Secondary Standard; Certified Reference Material
C10H8