EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A mesoporous-silica-immobilized oxovanadium cocatalyst for the lipase-catalyzed dynamic kinetic resolution of racemic alcohols.
PMID 23424181