EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Detection of mutations in the tyrosinase gene in a patient with type IA oculocutaneous albinism.
PMID 2342539
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3824
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid