EMAIL THIS PAGE TO A FRIEND
Lipids in health and disease
Butylated hydroxytoluene can protect polyunsaturated fatty acids in dried blood spots from degradation for up to 8 weeks at room temperature.
PMID 23425563
Abstract
Dried blood spots (DBS) from fingertip prick blood can enable high throughput fatty acid profiling but may be prone to lipid peroxidation during storage. The use of butylated hydroxytoluene (BHT) on chromatography paper can prevent polyunsaturated fatty acid (PUFA) loss but examinations on the length of storage times possible are not comprehensive. In the first study, venous whole blood was saturated on paper strips pre-soaked with 0, 2.5 or 5.0 mg/mL BHT and exposed to air for up to 28 days. In a second study, the effect of sealing DBS on 5.0 mg/mL BHT-soaked chromatography strips in capped test tubes or vacuum sealed polypropylene bags with and without nitrogen purging was examined over eight weeks. The fatty acid composition of the DBS were determined by gas chromatography and the effect of sample storage on omega-3 biomarkers were examined. PUFA and omega-3 biomarkers in DBS stored without BHT were dramatically reduced by day 3. In general, BHT delayed decreases in eicosapentaenoic + docosahexaenoic acid from baseline (3.2 ± 0.2 wt%) to 28 days (2.6 ± 0.03 wt%) of storage. In the % n-3 highly unsaturated fatty acids (HUFA) in total HUFA biomarker, BHT was more effective at preventing changes, particularly with 5.0 mg/mL BHT where no differences were detected up to 28 days. Sealed storage with BHT tended to increase the stability of the PUFA in DBS and nitrogen purging did not appear to provide additional benefits. The % n-3 HUFA in total HUFA biomarker also appeared to be more stable in the sealed storage study. The storage of DBS in sealed containers with BHT may prevent PUFA degradation for up to 8 weeks. The % n-3 HUFA in total HUFA biomarker appears to provide a more consistent assessment of omega-3 status throughout storage as compared with other omega-3 blood biomarkers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47168
3,5-Di-tert-4-butylhydroxytoluene (BHT), analytical standard