EMAIL THIS PAGE TO A FRIEND
Journal of visualized experiments : JoVE
RNA-seq analysis of transcriptomes in thrombin-treated and control human pulmonary microvascular endothelial cells.
PMID 23426025
Abstract
The characterization of gene expression in cells via measurement of mRNA levels is a useful tool in determining how the transcriptional machinery of the cell is affected by external signals (e.g. drug treatment), or how cells differ between a healthy state and a diseased state. With the advent and continuous refinement of next-generation DNA sequencing technology, RNA-sequencing (RNA-seq) has become an increasingly popular method of transcriptome analysis to catalog all species of transcripts, to determine the transcriptional structure of all expressed genes and to quantify the changing expression levels of the total set of transcripts in a given cell, tissue or organism. RNA-seq is gradually replacing DNA microarrays as a preferred method for transcriptome analysis because it has the advantages of profiling a complete transcriptome, providing a digital type datum (copy number of any transcript) and not relying on any known genomic sequence. Here, we present a complete and detailed protocol to apply RNA-seq to profile transcriptomes in human pulmonary microvascular endothelial cells with or without thrombin treatment. This protocol is based on our recent published study entitled "RNA-seq Reveals Novel Transcriptome of Genes and Their Isoforms in Human Pulmonary Microvascular Endothelial Cells Treated with Thrombin," in which we successfully performed the first complete transcriptome analysis of human pulmonary microvascular endothelial cells treated with thrombin using RNA-seq. It yielded unprecedented resources for further experimentation to gain insights into molecular mechanisms underlying thrombin-mediated endothelial dysfunction in the pathogenesis of inflammatory conditions, cancer, diabetes, and coronary heart disease, and provides potential new leads for therapeutic targets to those diseases. The descriptive text of this protocol is divided into four parts. The first part describes the treatment of human pulmonary microvascular endothelial cells with thrombin and RNA isolation, quality analysis and quantification. The second part describes library construction and sequencing. The third part describes the data analysis. The fourth part describes an RT-PCR validation assay. Representative results of several key steps are displayed. Useful tips or precautions to boost success in key steps are provided in the Discussion section. Although this protocol uses human pulmonary microvascular endothelial cells treated with thrombin, it can be generalized to profile transcriptomes in both mammalian and non-mammalian cells and in tissues treated with different stimuli or inhibitors, or to compare transcriptomes in cells or tissues between a healthy state and a disease state.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4648 Thrombin from bovine plasma, lyophilized powder, 40-300 NIH units/mg protein (biuret)
T7513 Thrombin from bovine plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
T6634 Thrombin from bovine plasma, lyophilized powder, 600-2,000 NIH units/mg protein (biuret)
T9549 Thrombin from bovine plasma, powder, cell culture tested, ≥1,500 NIH units/mg protein (E1%/280 = 19.5)
T7201 Thrombin from bovine plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
T7326 Thrombin from bovine plasma, ≥60 NIH units/mg protein (biuret)
T6884
Thrombin from human plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3)
T7009
Thrombin from human plasma, lyophilized powder, ≥1,000 NIH units/mg protein (E1%/280, 18.3)
T1063
Thrombin from human plasma, lyophilized powder, ≥2800 NIH units/mg protein (E1%/280, 18.3)
T8885
Thrombin from human plasma, lyophilized powder, Suitable for routine use in the thrombin time test
T4393
Thrombin from human plasma, lyophilized powder, 1500-3500 NIH units/mg protein (E1%/280, 18.3), cell culture tested
T7572
Thrombin from human plasma, 400-1000 NIH units/mg protein
T5772 Thrombin from rat plasma, lyophilized powder, ≥1,000 units/mg protein (biuret)
T9326
Thrombin human, BioUltra, recombinant, expressed in HEK 293 cells, aqueous solution, ≥95% (SDS-PAGE)