EMAIL THIS PAGE TO A FRIEND
The American journal of surgical pathology
Histologic evaluation of prophylactic hysterectomy and oophorectomy in Lynch syndrome.
PMID 23426126
Abstract
Women with Lynch syndrome (LS) are at increased risk for endometrial (EC) and ovarian carcinoma (OC). Current surveillance recommendations for detection of EC and OC in LS patients are not effective. Small studies have shown that prophylactic hysterectomy and bilateral salpingo-oophorectomy (P-TH-BSO) are the most effective and least expensive preventive measures in these patients. Data regarding histologic findings in prophylactic specimens in these patients are lacking. All LS patients who underwent P-TH-BSO at the Memorial Sloan-Kettering Cancer Center from 2000 to 2011 were identified. Slides were evaluated for the presence of endometrial hyperplasia (EH), EC, OC, or any other recurrent histologic findings. Twenty-five patients were identified, with an age range of 36 to 61 years. Fifteen patients had a synchronous or prior colorectal carcinoma, and 2 patients had a history of sebaceous carcinoma. Focal FIGO grade 1 endometrioid ECs were detected in 2 patients; 1 was 54 years of age (MSH2 mutation; superficially invasive), and the other was 56 years of age (MLH1 mutation; noninvasive). Focal complex atypical hyperplasia, unassociated with carcinoma, was seen in 3 patients, ages 35 and 45 (MLH1 mutations) and 53 years (MSH2 mutation). One patient (44 y, with MSH2 mutation) was found to have a mixed endometrioid/clear cell OC and simple EH without atypia. The OC was adherent to the colon but did not show distant metastasis. In our study, P-TH-BSOs performed because of the presence of LS revealed incidental EC and/or EH in 24% of cases and OC in 4%. The ECs were low grade, confined to the endometrium, and seen in patients older than 50 years. Prophylactic hysterectomy allows detection of early lesions in LS; these lesions appear to be small and focal. This small series of prophylactic hysterectomies may provide some clues about LS-associated endometrial carcinogenesis.